Merck
  • PTP1B is an effector of activin signaling and regulates neural specification of embryonic stem cells.
PTP1B is an effector of activin signaling and regulates neural specification of embryonic stem cells.
Cell stem cell (2013-10-22)
Kamil Matulka, Hsuan-Hwai Lin, Hana Hříbková, Dafe Uwanogho, Petr Dvořák, Yuh-Man Sun
ABSTRACT
During embryogenesis, the Activin/Nodal pathway promotes the mesendodermal lineage and inhibits neural fate. The molecular mechanisms underlying this role of the Activin/Nodal pathway are not clear. In this study, we report a role for protein tyrosine phosphatase 1B (PTP1B) in Activin-mediated early fate decisions during ESC differentiation and show that PTP1B acts as an effector of the Activin pathway to specify mesendodermal or neural fate. We found that the Activin/ALK4 pathway directly recruits PTP1B and stimulates its release from the endoplasmic reticulum through ALK4-mediated cleavage. Subsequently, PTP1B suppresses p-ERK1/2 signaling to inhibit neural specification and promote mesendodermal commitment. These findings suggest that a noncanonical Activin signaling pathway functions in lineage specification of mouse and human embryonic stem cells.
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
Anti-Oct-4 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse