Merck
  • Pembrolizumab in Combination with the Oncolytic Virus Pelareorep and Chemotherapy in Patients with Advanced Pancreatic Adenocarcinoma: A Phase Ib Study.
Pembrolizumab in Combination with the Oncolytic Virus Pelareorep and Chemotherapy in Patients with Advanced Pancreatic Adenocarcinoma: A Phase Ib Study.
Clinical cancer research : an official journal of the American Association for Cancer Research (2019-11-07)
Devalingam Mahalingam, Grey A Wilkinson, Kevin H Eng, Paul Fields, Patrick Raber, Jennifer L Moseley, Karol Cheetham, Matt Coffey, Gerard Nuovo, Pawel Kalinski, Bin Zhang, Sukeshi Patel Arora, Christos Fountzilas
ABSTRACT
Pelareorep is an intravenously delivered oncolytic reovirus that can induce a T-cell-inflamed phenotype in pancreatic ductal adenocarcinoma (PDAC). Tumor tissues from patients treated with pelareorep have shown reovirus replication, T-cell infiltration, and upregulation of PD-L1. We hypothesized that pelareorep in combination with pembrolizumab and chemotherapy in patients with PDAC would be safe and effective. A phase Ib single-arm study enrolled patients with PDAC who progressed after first-line treatment. Patients received pelareorep, pembrolizumab, and either 5-fluorouracil, gemcitabine, or irinotecan until disease progression or unacceptable toxicity. Study objectives included safety and dose-limiting toxicities, tumor response, evaluation for reovirus replication, and immune analysis in peripheral blood and tumor biopsies. Eleven patients were enrolled. Disease control was achieved in three of the 10 efficacy-evaluable patients. One patient achieved partial response for 17.4 months. Two additional patients achieved stable disease, lasting 9 and 4 months, respectively. Treatment was well tolerated, with mostly grade 1 or 2 treatment-related adverse events, including flu-like symptoms. Viral replication was observed in on-treatment tumor biopsies. T-cell receptor sequencing from peripheral blood revealed the creation of new T-cell clones during treatment. High peripheral clonality and changes in the expression of immune genes were observed in patients with clinical benefit. Pelareorep and pembrolizumab added to chemotherapy did not add significant toxicity and showed encouraging efficacy. Further evaluation of pelareorep and anti-PD-1 therapy is ongoing in follow-up studies. This research highlights the potential utility of several pretreatment and on-treatment biomarkers for pelareorep therapy warranting further investigation.
Product Description
Millipore
MILLIPLEX® Human TH17 Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple Th17 cytokine and chemokine biomarkers with the Th17 Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Human IL-18 Singleplex Magnetic Bead Kit