
Schisantherin A

Synonym(s):
Gomisin C, Schizantherin A, Wuweizi ester A
Empirical Formula (Hill Notation):
C30H32O9
CAS Number:
Molecular Weight:
536.57
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing