
Proxyphylline

Synonym(s):
7-(β-Hydroxypropyl)theophylline, (1,3-Dimethyl-7-[2-hydroxypropyl]-2,6-dioxopurine), Proxyphylline
Empirical Formula (Hill Notation):
C10H14N4O3
CAS Number:
Molecular Weight:
238.24
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing