
Photobiotin acetate salt

Synonym(s):
Biotin {3-[3-(4-azido-2-nitroanilino)-N-methylpropylamino]propylamide} acetate salt, N-(4-Azido-2-nitrophenyl)-N′-(3-biotinylaminopropyl)-N′-methyl-1,3-propanediamine acetate salt
Empirical Formula (Hill Notation):
C23H35N9O4S · C2H4O2
CAS Number:
Molecular Weight:
593.70
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing