
Pergolide mesylate

Synonym(s):
Pergolide mesylate salt, 8β-[(Methylthio)methyl]-6-propylergoline methanesulfonate salt, Pergolide methanesulfonate salt
Empirical Formula (Hill Notation):
C19H26N2S · CH4O3S
CAS Number:
Molecular Weight:
410.59
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing