
Melamine-13C3

Synonym(s):
1,3,5-Triazine-2,4,6-triamine-13C3
Empirical Formula (Hill Notation):
13C3H6N6
CAS Number:
Molecular Weight:
129.10
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing