Skip to Content
Merck
Ibuprofen Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material

Ibuprofen Related Compound C

Ibuprofen Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
4′-Isobutylacetophenone, 1-(4-Isobutylphenyl)ethanone, 1-Acetyl-4-isobutylbenzene
Empirical Formula (Hill Notation):
C12H16O
CAS Number:
Molecular Weight:
176.25
EC Number:

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique