
Genkwanin

Synonym(s):
4′,5-Dihydroxy-7-methoxyflavone, 5-Hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-1-benzopyran-4-one, 7-O-Methylapigenin, Apigenin 7-O-methyl ether, Puddumetin
Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing