
Difenoconazol

Empirical Formula (Hill Notation):
C19H17Cl2N3O3
CAS Number:
Molecular Weight:
406.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2