
Clopidogrel bisulfate

Synonym(s):
(S)-Methyl (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogensulfate
Empirical Formula (Hill Notation):
C16H16ClNO2S · H2SO4
CAS Number:
Molecular Weight:
419.90
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2