
3,5-Di-caffeoylquinic acid

Synonym(s):
3,5-Di-caffeoylquinic acid
Empirical Formula (Hill Notation):
C25H24O12
CAS Number:
Molecular Weight:
516.45
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing