
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphoethanolamine

Synonym(s):
1-Hexadecanoyl-2-(cis-9-octadecenoyl)-sn-glycero-3-phosphoethanolamine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine, 3-sn-Phosphatidylethanolamine, 2-Oleoyl-1-palmitoyl, L-β-Oleoyl-γ-palmitoyl-α-cephalin, POPE
Linear Formula:
C39H76NO8P
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing