
2,4-Dimethyl-5-acetylthiazole

Synonym(s):
5-Acetyl-2,4-dimethylthiazole
Empirical Formula (Hill Notation):
C7H9NOS
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing