Skip to Content
Merck
All Photos(2)

Key Documents

SML3734

Sigma-Aldrich

AO-159

≥98% (HPLC)

Synonym(s):

2,3,6,7-Tetramethyl 9-(4-methoxyphenyl)-1,4,8-trithiaspiro[4.5]deca-2,6,9-triene-2,3,6,7-tetracarboxylate, 9-(4-Methoxyphenyl)-1,4,8-trithiaspiro[4.5]deca-2,6,9-triene-2,3,6,7-tetracarboxylic acid 2,3,6,7-tetramethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20O9S3
CAS Number:
Molecular Weight:
524.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

AO-159 is a cell penetrant and potent pan-HMT (pan- Histone Methyltransferase) inhibitor that effectively inhibit the enzymatic activity of both G9a and Suv39H1 enzymes. AO-159 potently reduces the levels of H3K9me1, H3K9me2, and H3K9me3 in COS7 cells. In inhibits the five histone H3K9 methylating enzymes Suv39H1, Suv39H2, SETDB1, G9a, and GLP at low micromolar concentrations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visualization of the dynamic interaction between nucleosomal histone H3K9 tri-methylation and HP1? chromodomain in living cells
Cell Chemical Biology, 29(7), 1153-1161 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service