Skip to Content
Merck
All Photos(1)

Key Documents

EP0030120191

Eppendorf® Safe-Lock micro test tubes

capacity 1.5 mL, amber, pack of 1000 ea

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.15

material

amber
amber cap
amber tube
cap (plug seal)
conical bottom
polypropylene cap
polypropylene tube

feature

graduations

packaging

pack of 1000 ea

manufacturer/tradename

Eppendorf® 0030120191

capacity

1.5 mL

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • The hinged Eppendorf® Safe-Lock lid provides additional protection against unintentional lid opening during incubation and storage.
  • g-Safe®:Provides great centrifugation stability and prevents sample loss due to tube breakage
  • The optimized sealing properties ensures safety when working with hazardous samples and minimal evaporation rates during long-term storage.
  • The large frosted lid and wide side-surface area make labeling convenient.
  • Functions between temperatures -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
  • The polypropylene protects against chemical and mechanical stress, and extreme temperature.

Other Notes

Manufactured without slip agents, plasticizers, and biocides substances that negatively affect bio-assay results.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service