Merck
HomeADC & BioconjugationCrosslinkers Selection Guide
Crosslinkers Selection Guide
Searching for the perfect crosslinker for your bioconjugation research? Explore our detailed selection guide, conveniently organized by crosslinker type, reactive groups, spacer arm length, and more, to assist you in making the right choice.
Section Overview
The graphic depicts a biological process involving proteins. On the left, two starburst formations, one red and one purple, are labeled "Protein-Protein Interaction," showing that proteins interact. To their right is a blue squiggly line labeled "Crosslinker." The next symbol is a purple circle with squiggly lines ringed by a purple arrow, which represents bioconjugation. On the far right, two starburst structures are connected by a blue squiggly line labeled "Crosslinked Proteins."
A crosslinker converts interacting proteins into crosslinked proteins via bioconjugation.
WHAT ARE CROSSLINKERS
In chemical biology and bioconjugation, crosslinkers are essential chemical compounds that connect two or more molecules by forming covalent bonds. These linkers are pivotal in numerous applications, including the creation of bioconjugates, the exploration of protein-protein interactions, and the development of biomaterials. Crosslinkers come in various types, each with specific functionalities tailored to different applications.
Crosslinker Selection Reference Table
Crosslinker
Reactive Groups
Spacer Arm Length
Water Solubility
Cleavability
Crosslinking Efficiency
Stability
Toxicity/Biocompatibility
DSS
NHS ester (amine)
11.4 Å (rigid)
Low
Non-cleavable
High
Stable, sensitive to moisture
Moderate toxicity
Sulfo-SMCC
NHS ester (amine), maleimide (thiol)
11.6 Å (rigid)
High
Non-cleavable
High
Stable under physiological conditions, light-sensitive
Low toxicity, in vivo suitable
EDC
Carbodiimide (carboxyl/amine)
Zero-length
High
Cleavable under acidic conditions
High
Stable under acidic conditions
Low toxicity, in vivo suitable
BS3
NHS ester (amine)
11.4 Å (rigid)
High
Non-cleavable
High
Stable, sensitive to moisture
Moderate toxicity
DTSSP
NHS ester (amine), disulfide (thiol)
12.0 Å (rigid)
High
Cleavable under reducing conditions
High
Stable, reducing agents cleave
Low toxicity, in vivo suitable
Sulfo-NHS-LC-Biotin
NHS ester (amine)
22.4 Å (flexible)
High
Non-cleavable
High
Stable, moisture sensitive
Low toxicity
BMH
Maleimide (thiol)
16.1 Å (flexible)
Low
Non-cleavable
High
Stable, light-sensitive
Moderate toxicity
DMP
NHS ester (amine)
9.2 Å (rigid)
Low
Non-cleavable
Moderate
Stable
Moderate toxicity
MBS
NHS ester (amine), maleimide (thiol)
10.2 Å (rigid)
Low
Non-cleavable
High
Stable under physiological conditions, light-sensitive
Moderate toxicity
PEGylated Crosslinkers
NHS ester (amine)
Variable (depends on PEG length)
High
Cleavable and non-cleavable variants
High
Stable, customizable based on PEG length
Low toxicity, in vivo suitable
Notes:
  • DSS (Disuccinimidyl suberate) is a popular homo-bifunctional crosslinker with high reactivity but moderate toxicity.
  • Sulfo-SMCC is a widely used hetero-bifunctional crosslinker with high solubility and low toxicity, suitable for in vivo applications.
  • EDC (1-ethyl-3-(3-dimethylaminopropyl)carbodiimide) is a zero-length crosslinker known for its high efficiency and low toxicity.
  • BS3 (Bis[sulfosuccinimidyl] suberate) is similar to DSS but with higher water solubility.
  • DTSSP (3,3'-Dithiobis[sulfosuccinimidylpropionate]) offers cleavability under reducing conditions.
  • Sulfo-NHS-LC-Biotin is used for biotinylation with a flexible spacer arm.
  • BMH (Bismaleimidohexane) provides flexibility with maleimide groups for thiol reactivity.
  • DMP (Dimethyl pimelimidate) is less commonly used but effective for specific applications.
  • MBS (m-Maleimidobenzoyl-N-hydroxysuccinimide ester) is another hetero-bifunctional option with specific applications.
  • PEGylated Crosslinkers offer customizable spacer lengths and are highly biocompatible.
How Do Crosslinkers Work?
Crosslinkers operate by reacting with specific functional groups on molecules. Amine-reactive groups, such as NHS esters and isocyanates, react with primary amines on proteins or other biomolecules. Sulfhydryl-reactive groups, including maleimides and pyridyl disulfides, react with thiol groups on cysteine residues, while carboxyl-reactive groups, like carbodiimides, activate carboxyl groups to form amide bonds with amines.
WHERE CAN CROSSLINKERS BE USED?
The applications of crosslinkers are broad and varied. They are instrumental in protein-protein interaction studies by stabilizing transient interactions between proteins, which can then be analyzed using techniques like mass spectrometry or Western blotting. In bioconjugation, crosslinkers attach drugs, fluorescent labels, or other probes to biomolecules, facilitating therapeutic development and diagnostics. In material science, crosslinkers help create networks in hydrogels and other biomaterials that can mimic biological tissues. Additionally, they enable the immobilization of biomolecules, such as enzymes or antibodies, on solid supports, enhancing stability and reusability in various assays.
Homo-Bifunctional vs. Hetero-Bifunctional Crosslinkers
Homo-bifunctional crosslinkers, which possess identical reactive groups at both ends, are used to link similar molecules, such as connecting two proteins that share the same functional group. An example of this type is disuccinimidyl suberate (DSS), which features two N-hydroxysuccinimide (NHS) ester groups. In contrast, hetero-bifunctional crosslinkers have different reactive groups at each end, making them ideal for linking diverse molecules, like a protein and a small molecule, each reacting with a different functional group. Sulfo-SMCC, with an NHS ester on one end and a maleimide group on the other, exemplifies this type.
SPECIAL CONSIDERATIONS WHEN CHOOSING CROSSLINKERS
Choosing the right crosslinker involves several key considerations. Specificity is crucial, as the crosslinker must target specific functional groups on the molecules of interest. The length of the spacer arm in the crosslinker affects the distance between linked molecules, influencing the function and stability of the conjugate. Water-soluble crosslinkers are preferred for biological applications to ensure compatibility with aqueous environments. Furthermore, some crosslinkers are designed to be cleavable under certain conditions, allowing for the controlled release of linked molecules.
Related Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch