HPLC Analysis of Warfarin Enantiomers on Astec® CYCLOBOND I 2000 (UV at 278 nm)
Materials
analytical column
Product No.
Description
Pricing
Application
Product No.
Description
Pricing
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V (Method 1)
application for HPLCHPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V (Method 2)
application for HPLCCONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] acetonitrile; [B] acetic acid; [C] TEA; (100:0.3:0.2, A:B:C)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 278 nm
injection
2 μL
sample
2 mg/mL in mobile phase
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null
null