HPLC Analysis of Warfarin Anticoagulant on Ascentis® C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis C18, 15 cm x 4.6 mm I.D., 5 μm particles (581324-U)
mobile phase
[A] 20 mM phosphoric acid (pH 2.0, unadjusted): [B] methanol; (30:70, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 280 nm
injection
10 μL
sample
50 μg/mL each in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany