- FixK
FixK
Frontiers in microbiology (2017-09-16)
María J Torres, Emilio Bueno, Andrea Jiménez-Leiva, Juan J Cabrera, Eulogio J Bedmar, Socorro Mesa, María J Delgado
PMID28912756
ABSTRACT
The powerful greenhouse gas, nitrous oxide (N
Sigma-Aldrich
Anti-Sheep IgG (whole molecule)–Peroxidase antibody produced in donkey, affinity isolated antibody, buffered aqueous solution