Merck
  • Dynamic Regulation of Hypothalamic DMXL2, KISS1, and RFRP Expression During Postnatal Development in Non-Human Primates.
Dynamic Regulation of Hypothalamic DMXL2, KISS1, and RFRP Expression During Postnatal Development in Non-Human Primates.
Molecular neurobiology (2016-12-14)
Fazal Wahab, Charis Drummer, Stefan Schlatt, Rüdiger Behr
ABSTRACT
The neurobiological mechanism of puberty onset in primates is currently only partly understood. A recent study reported an important role of Dmx-like 2 (DMXL2), a gene encoding rabconnectin-3α vesicular protein, in human subjects with mental retardation and neuroendocrine impairment of reproduction. To further characterize the potential role of DMXL2 in the regulation of reproduction, we analyzed the expression of DMXL2 in hypothalami of newborn, infantile, juvenile, pubertal, and postpubertal female and male common marmoset monkeys. Additionally, as the relative hypothalamic levels of gonadotropin-inhibitory hormone (GnIH) transcript during postnatal development are unknown in primates, we also quantified messenger RNA (mRNA) levels of RFRP, a gene encoding GnIH. Moreover, the transcript levels of kisspeptin, a well-known regulator of the hypothalamic neurohormonal axis controlling reproduction, were also checked. Transcript and protein levels of DMXL2 and Kiss1 transcript levels increase from the newborn to the infantile and from the juvenile (prepubertal) to the pubertal and the postpubertal period. We also noted a clear upsurge in RFRP transcript levels in the prepubertal period. In conclusion, the hypothalamic expressions of Kiss1 and DMXL2 mRNA increase during infantile, pubertal, and adult stages compared to newborn and juvenile stages in common marmoset monkeys. In contrast, the expression of RFRP mRNA upsurges in juvenile monkeys. Further mechanistic studies are needed to characterize the potential inhibitory role of the GnIH-GPR147 signaling in the prepubertal period and the role of DMXL2 in the molecular cascade regulating the neuroendocrine reproductive axis in primates.
Sigma-Aldrich
Anti-DMXL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.