Merck
  • A cryptic Tudor domain links BRWD2/PHIP to COMPASS-mediated histone H3K4 methylation.
A cryptic Tudor domain links BRWD2/PHIP to COMPASS-mediated histone H3K4 methylation.
Genes & development (2017-11-02)
Marc A J Morgan, Ryan A Rickels, Clayton K Collings, Xiaolin He, Kaixiang Cao, Hans-Martin Herz, Kira A Cozzolino, Nebiyu A Abshiru, Stacy A Marshall, Emily J Rendleman, Christie C Sze, Andrea Piunti, Neil L Kelleher, Jeffrey N Savas, Ali Shilatifard
ABSTRACT
Histone H3 Lys4 (H3K4) methylation is a chromatin feature enriched at gene
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 486.7 mPa.s (25 °C)