Merck
  • Improved surface-enhanced Raman and catalytic activities of reduced graphene oxide-osmium hybrid nano thin films.
Improved surface-enhanced Raman and catalytic activities of reduced graphene oxide-osmium hybrid nano thin films.
Royal Society open science (2017-10-11)
C Kavitha, K Bramhaiah, Neena S John, Shantanu Aggarwal
ABSTRACT
Reduced graphene oxide-osmium (rGO-Os) hybrid nano dendtrites have been prepared by simple liquid/liquid interface method for the first time. The method involves the introduction of phase-transfered metal organic precursor in toluene phase and GO dispersion in the aqueous phase along with hydrazine hydrate as the reducing agent. Dendritic networks of Os nanoparticles and their aggregates decorating rGO layers are obtained. The substrate shows improved catalytic and surface-enhanced activities comparable with previous reports. The catalytic activity was tested for the reduction of
Sigma-Aldrich
Osmium(III) chloride, 99.9% trace metals basis
Pricing and availability is not currently available.