Merck
  • Plasma and erythrocyte uptake of omega-3 fatty acids from an intravenous fish oil based lipid emulsion in patients with advanced oesophagogastric cancer.
Plasma and erythrocyte uptake of omega-3 fatty acids from an intravenous fish oil based lipid emulsion in patients with advanced oesophagogastric cancer.
Clinical nutrition (Edinburgh, Scotland) (2016-06-28)
A M Eltweri, A L Thomas, H L Fisk, A Arshad, P C Calder, A R Dennison, D J Bowrey
ABSTRACT
It has been demonstrated that short term intravenous (IV) administration of omega-3 polyunsaturated fatty acids (PUFAs) is more effective than oral supplementation at promoting incorporation of the bioactive omega-3 PUFAs eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA) into plasma, blood cells and tissues. The effect of repeated short term IV infusion of omega-3 PUFAs was investigated in patients with advanced oesophagogastric cancer during palliative chemotherapy. Patients with advanced oesophagogastric cancer (n = 21) were recruited into a phase II pilot clinical trial. All patients were scheduled for an intravenous infusion of Omegaven Twenty patients received at least one Omegaven Repeated weekly omega-3 PUFA infusion is effective in enriching plasma PC and RBC membranes in EPA in patients with advanced oesophagogastric cancer receiving palliative chemotherapy. Clinical Trials.Gov NCT01870791.
Sigma-Aldrich
all-cis-7,10,13,16,19-Docosapentaenoic acid, synthetic, ≥97%