Merck
  • Hydrogen sulfide protects against endoplasmic reticulum stress and mitochondrial injury in nucleus pulposus cells and ameliorates intervertebral disc degeneration.
Hydrogen sulfide protects against endoplasmic reticulum stress and mitochondrial injury in nucleus pulposus cells and ameliorates intervertebral disc degeneration.
Pharmacological research (2017-01-15)
Daoliang Xu, Haiming Jin, Jianxia Wen, Jiaoxiang Chen, Deheng Chen, Ningyu Cai, Yongli Wang, Jianle Wang, Yu Chen, Xiaolei Zhang, Xiangyang Wang
ABSTRACT
It has been suggested that excessive apoptosis in intervertebral disc cells induced by inflammatory cytokines, such as interleukin (IL)-1β, is related to the process of intervertebral disc degeneration (IVDD). Hydrogen sulfide (H
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
MISSION® esiRNA, targeting human CTH