Merck
  • Organic cation transporter 1 (OCT1/mOct1) is localized in the apical membrane of Caco-2 cell monolayers and enterocytes.
Organic cation transporter 1 (OCT1/mOct1) is localized in the apical membrane of Caco-2 cell monolayers and enterocytes.
Molecular pharmacology (2013-05-18)
Tianxiang Kevin Han, Ruth S Everett, William R Proctor, Chee M Ng, Chester L Costales, Kim L R Brouwer, Dhiren R Thakker
ABSTRACT
Organic cation transporters (OCTs) are members of the solute carrier 22 family of transporter proteins that are involved in absorption, distribution, and excretion of organic cations. OCT3 is localized in the apical (AP) membrane of enterocytes, but the literature is ambiguous about OCT1 (mOct1) localization, with some evidence suggesting a basolateral (BL) localization in human and mouse enterocytes. This is contrary to our preliminary findings showing AP localization of OCT1 in Caco-2 cell monolayers, an established model of human intestinal epithelium. Therefore, this study aims at determining the localization of OCT1 (mOct1) in Caco-2 cells, and human and mouse enterocytes. Functional studies using OCT1-specific substrate pentamidine showed transporter-mediated AP but not BL uptake in Caco-2 cells and human and mouse intestinal tissues. OCT1 inhibition decreased AP uptake of pentamidine by ∼50% in all three systems with no effect on BL uptake. A short hairpin RNA-mediated OCT1 knockdown in Caco-2 cells decreased AP uptake of pentamidine by ∼50% but did not alter BL uptake. Immunostaining and confocal microscopy in all three systems confirmed AP localization of OCT1 (mOct1). Our studies unequivocally show AP membrane localization of OCT1 (mOct1) in Caco-2 cells and human and mouse intestine. These results are highly significant as they will require reinterpretation of previous drug disposition and drug-drug interaction studies where conclusions were drawn assuming BL localization of OCT1 in enterocytes. Most importantly, these results will require revision of the regulatory guidance for industry in the United States and elsewhere because it has stated that OCT1 is basolaterally localized in enterocytes.
Sigma-Aldrich
Anti-SLC22A1 (AB1) antibody produced in rabbit, IgG fraction of antiserum