Merck
  • Modulation of the dystrophin-associated protein complex in response to resistance training in young and older men.
Modulation of the dystrophin-associated protein complex in response to resistance training in young and older men.
Journal of applied physiology (Bethesda, Md. : 1985) (2008-03-22)
David J Kosek, Marcas M Bamman
ABSTRACT
The dystrophin-associated protein complex (DAPC) is a scaffold of proteins linking the intracellular cytoskeleton with the extracellular matrix that is integral to structural stability and integrity, signaling and mechanotransduction, and force transmission. We hypothesized that the expression of DAPC component proteins would be altered by resistance loading during progressive resistance training (PRT)-mediated myofiber hypertrophy, and we investigated whether aging influenced these changes. Seventeen young (27 yr) and 13 older (65 yr) men completed 16 wk of PRT with muscle biopsies at baseline (T1), 24 h after bout 1 (T2), and 24 h after the final bout at week 16 (T3). Myofiber hypertrophy in the young (type I 31%, P < 0.005; type II 40%, P < 0.001) far exceeded hypertrophy in the old (type II only, 19.5%, P < 0.05). PRT altered protein expression for caveolin-3 (decreased 24% by T3, P < 0.01), alpha(1)-syntrophin (increased 16% by T3, P < 0.05), alpha-dystrobrevin (fell 23% from T2 to T3, P < 0.01), and dystrophin [rose acutely (30% by T2, P < 0.05) and returned to baseline by T3]. The phosphorylation state of membrane neuronal nitric oxide synthase (Ser(1417)) decreased 70% (P < 0.005) by T3, particularly in the old (81%), whereas p38 MAPK phosphorylation increased twofold by T3 in the old (P < 0.01). We conclude that component proteins of the DAPC are modulated by PRT, which may serve to improve both structural and signaling functions during load-mediated myofiber hypertrophy. The blunted hypertrophic adaptation seen in old vs. young men may have resulted from overstress, as suggested by marked p38 MAPK activation in old men only.
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-phospho-nNOS/NOS I (Ser1417) Antibody, Upstate®, from rabbit