Merck
Epigenetic roles of MLL oncoproteins are dependent on NF-κB.
Cancer cell (2013-09-24)
Hsu-Ping Kuo, Zhong Wang, Dung-Fang Lee, Masayuki Iwasaki, Jesus Duque-Afonso, Stephen H K Wong, Chiou-Hong Lin, Maria E Figueroa, Jie Su, Ihor R Lemischka, Michael L Cleary
ABSTRACT
MLL fusion proteins in leukemia induce aberrant transcriptional elongation and associated chromatin perturbations; however, the upstream signaling pathways and activators that recruit or retain MLL oncoproteins at initiated promoters are unknown. Through functional and comparative genomic studies, we identified an essential role for NF-κB signaling in MLL leukemia. Suppression of NF-κB led to robust antileukemia effects that phenocopied loss of functional MLL oncoprotein or associated epigenetic cofactors. The NF-κB subunit RELA occupies promoter regions of crucial MLL target genes and sustains the MLL-dependent leukemia stem cell program. IKK/NF-κB signaling is required for wild-type and fusion MLL protein retention and maintenance of associated histone modifications, providing a molecular rationale for enhanced efficacy in therapeutic targeting of this pathway in MLL leukemias.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IKK Inhibitor III, BMS-345541, IKK Inhibitor III, BMS-345541, CAS 445430-58-0, is a cell-permeable, potent, selective, and allosteric site-binding inhibitor of IKK-2 (IC₅₀ = ~ 300 nM).
Sigma-Aldrich
IKK Inhibitor VII, The IKK Inhibitor VII, also referenced under CAS 873225-46-8, controls the biological activity of IKK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
IKK-2 Inhibitor IV, The IKK-2 Inhibitor IV, also referenced under CAS 507475-17-4, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.