Merck
  • 24(S)-Hydroxycholesterol as a Modulator of Neuronal Signaling and Survival.
24(S)-Hydroxycholesterol as a Modulator of Neuronal Signaling and Survival.
The Neuroscientist : a review journal bringing neurobiology, neurology and psychiatry (2015-01-30)
Min-Yu Sun, Andrew J Linsenbardt, Christine M Emnett, Lawrence N Eisenman, Yukitoshi Izumi, Charles F Zorumski, Steve Mennerick
ABSTRACT
The major cholesterol metabolite in brain, 24(S)-hydroxycholesterol (24S-HC), serves as a vehicle for cholesterol removal. Its effects on neuronal function, however, have only recently begun to be investigated. Here, we review that nascent work. Our own studies have demonstrated that 24S-HC has potent positive modulatory effects on N-methyl-d-aspartate (NMDA) receptor (NMDAR) function. This could have implications not only for brain plasticity but also for pathological NMDAR overuse. Other work has demonstrated effects of 24S-HC on neuronal survival and as a possible biomarker of neurodegenerative disease. Depending on circumstances, both upregulation/mimicry of 24S-HC signaling and down-regulation/antagonism may have therapeutic potential. We are interested in the possibility that synthetic analogues of 24S-HC with positive effects at NMDARs may hold neurotherapeutic promise, given the role of NMDA receptor hypofunction in certain neuropsychiatric disorders.
Sigma-Aldrich
24(S)-Hydroxycholesterol, ≥98% (HPLC)