Merck
  • Identification of a critical determinant that enables efficient fatty acid synthesis in oleaginous fungi.
Identification of a critical determinant that enables efficient fatty acid synthesis in oleaginous fungi.
Scientific reports (2015-06-11)
Haiqin Chen, Guangfei Hao, Lei Wang, Hongchao Wang, Zhennan Gu, Liming Liu, Hao Zhang, Wei Chen, Yong Q Chen
ABSTRACT
Microorganisms are valuable resources for lipid production. What makes one microbe but not the other able to efficiently synthesize and accumulate lipids is poorly understood. In the present study, global gene expression prior to and after the onset of lipogenesis was determined by transcriptomics using the oleaginous fungus Mortierella alpina as a model system. A core of 23 lipogenesis associated genes was identified and their expression patterns shared a high similarity among oleaginous microbes Chlamydomonas reinhardtii, Mucor circinelloides and Rhizopus oryzae but was dissimilar to the non-oleaginous Aspergillus nidulans. Unexpectedly, Glucose-6-phosphate dehydrogenase (G6PD) and 6-phosphogluconate dehydrogenase (PGD) in the pentose phosphate pathway (PPP) were found to be the NADPH producers responding to lipogenesis in the oleaginous microbes. Their role in lipogenesis was confirmed by a knockdown experiment. Our results demonstrate, for the first time, that the PPP plays a significant role during fungal lipogenesis. Up-regulation of NADPH production by the PPP, especially G6PD, may be one of the critical determinants that enables efficiently fatty acid synthesis in oleaginous microbes.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate