Merck
  • Ryanodine receptor 2 contributes to hemorrhagic shock-induced bi-phasic vascular reactivity in rats.
Ryanodine receptor 2 contributes to hemorrhagic shock-induced bi-phasic vascular reactivity in rats.
Acta pharmacologica Sinica (2014-09-30)
Rong Zhou, Xiao-li Ding, Liang-ming Liu
ABSTRACT
Ryanodine receptor 2 (RyR2) is a critical component of intracellular Ca(2+) signaling in vascular smooth muscle cells (VSMCs). The aim of this study was to investigate the role of RyR2 in abnormal vascular reactivity after hemorrhagic shock in rats. SD rats were hemorrhaged and maintained mean arterial pressure (MAP) at 40 mmHg for 30 min or 2 h, and then superior mesenteric arteries (SMA) rings were prepared to measure the vascular reactivity. In other experiments, SMA rings of normal rats and rat VSMCs were exposed to a hypoxic medium for 10 min or 3 h. SMA rings of normal rats and VSMCs were transfected with siRNA against RyR2. Intracellular Ca(2+) release in VSMCs was assessed using Fura-2/AM. The vascular reactivity of the SMA rings from hemorrhagic rats was significantly increased in the early stage (30 min), but decreased in the late stage (2 h) of hemorrhagic shock. Similar results were observed in the SMA rings exposed to hypoxia for 10 min or 3 h. The enhanced vascular reactivity of the SMA rings exposed to hypoxia for 10 min was partly attenuated by transfection with RyR2 siRNA, whereas the blunted vascular reactivity of the SMA rings exposed to hypoxia for 3 h was partly restored by transfection with RyR2 siRNA. Treatment with the RyR agonist caffeine (1 mmol/L) significantly increased Ca(2+) release in VSMCs exposed to hypoxia for 10 min or 3 h, which was partially antagonized by transfection with RyR2 siRNA. RyR2-mediated Ca(2+) release contributes to the development of bi-phasic vascular reactivity induced by hemorrhagic shock or hypoxia.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cd82
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rrm2
Sigma-Aldrich
MISSION® esiRNA, targeting human CD1E
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human RRM2
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous