Merck
  • Method validation for determination of alkaloid content in goldenseal root powder.
Method validation for determination of alkaloid content in goldenseal root powder.
Journal of AOAC International (2003-07-11)
Holly A Weber, Matthew K Zart, Andrew E Hodges, Kellie D White, Sarah M Barnes, Leslie A Moody, Alice P Clark, Roger K Harris, J Diane Overstreet, Cynthia S Smith
ABSTRACT
A fast, practical ambient extraction methodology followed by isocratic liquid chromatography (LC) analysis with UV detection was validated for the determination of berberine, hydrastine, and canadine in goldenseal (Hydrastis canadensis L.) root powder. The method was also validated for palmatine, a major alkaloid present in the possible bioadulterants Coptis, Oregon grape root, and barberry bark. Alkaloid standard solutions were linear over the evaluated concentration ranges. The analytical method was linear for alkaloid extraction using 0.3-2 g goldenseal root powder/100 mL extraction solvent. Precision of the method was demonstrated using 10 replicate extractions of 0.5 g goldenseal root powder, with percent relative standard deviation for all 4 alkaloids < or = 1.6. Alkaloid recovery was determined by spiking each alkaloid into triplicate aliquots of neat goldenseal root powder. Recoveries ranged from 92.3% for palmatine to 101.9% for hydrastine. Ruggedness of the method was evaluated by performing multiple analyses of goldenseal root powder from 3 suppliers over a 2-year period. The method was also used to analyze Coptis root, Oregon grape root, barberry bark, and celandine herb, which are possible goldenseal bioadulterants. The resulting chromatographic profiles of the bioadulterants were significantly different from that of goldenseal. The method was directly transferred to LC with mass spectrometry, which was used to confirm the presence of goldenseal alkaloids tetrahydroberberastine, berberastine, canadaline, berberine, hydrastine, and canadine, as well as alkaloids from the bioadulterants, including palmatine, jatrorrhizine, and coptisine.
Sigma-Aldrich
Palmatine chloride hydrate, 97%