Merck
  • Utility of rigid bronchoscopic dilatation and mitomycin C application in the management of postintubation tracheal stenosis: case series and systematic review of literature.
Utility of rigid bronchoscopic dilatation and mitomycin C application in the management of postintubation tracheal stenosis: case series and systematic review of literature.
Journal of bronchology & interventional pulmonology (2012-12-05)
Karan Madan, Ritesh Agarwal, Ashutosh N Aggarwal, Dheeraj Gupta
ABSTRACT
Postintubation tracheal stenosis (PITS) is a common problem encountered by interventional pulmonologists. The aim of this study was to evaluate the utility of mitomycin C (MMC) as an adjunctive treatment to rigid bronchoscopic dilatation in patients with PITS. Prospective analysis of data from the interventional pulmonology unit of a large tertiary care teaching center in North India. Patients with a diagnosis of PITS undergoing rigid bronchoscopic dilatation and MMC (0.4 mg/mL) application were included. The primary outcome was the occurrence of restenosis. Seven patients underwent rigid bronchoscopic dilatation, followed by the application of MMC at 4 quadrants of the stenosis. Controlled radial expansion balloon bronchoplasty was also performed, if necessary, in addition to mechanical dilatation using the barrel of the rigid bronchoscope. Restenosis occurred in all 7 patients (100%) and the mean duration to the detection of restenosis was 27 days. The restenosis was symptomatic in 6 out of 7 (85.7%) patients. Rigid bronchoscopic dilatation and a single application of MMC is not an effective treatment in the management of PITS.
Product Description
Mitomycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture