Merck
  • Antibiotics for gram-positive bacterial infections: vancomycin, teicoplanin, quinupristin/dalfopristin, oxazolidinones, daptomycin, dalbavancin, and telavancin.
Antibiotics for gram-positive bacterial infections: vancomycin, teicoplanin, quinupristin/dalfopristin, oxazolidinones, daptomycin, dalbavancin, and telavancin.
Infectious disease clinics of North America (2009-11-17)
Michael D Nailor, Jack D Sobel
ABSTRACT
An overview of the mechanism of action, dosing, clinical indications, and toxicities of the glycopeptide vancomycin is provided. The emerging gram-positive bacterial resistance to antimicrobials and its mechanisms are reviewed. Strategies to control this emergence of resistance are expected to be proposed. Newer antimicrobial agents that have activity against vancomycin-resistant organisms are now available and play a critical role in the treatment of life-threatening infections.
Product Description
Teicoplanin for identification, European Pharmacopoeia (EP) Reference Standard
Teicoplanin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Teicoplanin