Merck
  • Production, characterization, and identification using proteomic tools of a polygalacturonase from Fusarium graminearum.
Production, characterization, and identification using proteomic tools of a polygalacturonase from Fusarium graminearum.
Journal of basic microbiology (2014-01-10)
Leonel M Ortega, Gisele E Kikot, Natalia L Rojas, Laura M I López, Andrea L Astoreca, Teresa M Alconada
ABSTRACT
Since enzymatic degradation is a mechanism or component of the aggressiveness of a pathogen, enzymatic activities from a Fusarium graminearum isolate obtained from infected wheat spikes of Argentina Pampa region were studied in order to understand the disease progression, tending to help disease control. In particular, the significance of the study of polygalacturonase activity is based on that such activity is produced in the early stages of infection on the host, suggesting a crucial role in the establishment of disease. In this sense, polygalacturonase activity produced by this microorganism has been purified 375 times from 2-day-old culture filtrates by gel filtration and ion-exchange chromatography successively. The purified sample showed two protein bands in sodium dodecyl sulfate-polyacrylamide gels, with a molecular mass of 40 and 55 kDa. The protein bands were identified as an endopolygalacturonase and as a serine carboxypeptidase of F. graminearum, respectively, by peptide mass fingerprinting (matrix-assisted laser desorption/ionization time-of-flight (MALDI TOF/TOF) fragment ion analysis). The pattern of substrate degradation analyzed by thin layer chromatography confirmed the mode of action of the enzyme as an endopolygalacturonase. High activity of the polygalacturonase against polygalacturonic acid was observed between 4 and 6 of pH, and between 30 and 50 °C, being 5 and 50 °C the optimum pH and temperature, respectively. The enzyme was fully stable at pH 5 for 120 min and 30 °C and sensible to the presence of some metal ions. This information would contribute to understand the most favorable environmental conditions for establishment of the disease.
Sigma-Aldrich
Digalacturonic acid, ≥85% (HPLC)