Merck
  • Characterization of the new AmpC β-lactamase FOX-8 reveals a single mutation, Phe313Leu, located in the R2 loop that affects ceftazidime hydrolysis.
Characterization of the new AmpC β-lactamase FOX-8 reveals a single mutation, Phe313Leu, located in the R2 loop that affects ceftazidime hydrolysis.
Antimicrobial agents and chemotherapy (2013-07-24)
Francisco José Pérez-Llarena, Frédéric Kerff, Laura Zamorano, María Carmen Fernández, Maria Luz Nuñez, Elisenda Miró, Antonio Oliver, Ferran Navarro, Germán Bou
ABSTRACT
A novel class C β-lactamase (FOX-8) was isolated from a clinical strain of Escherichia coli. The FOX-8 enzyme possessed a unique substitution (Phe313Leu) compared to FOX-3. Isogenic E. coli strains carrying FOX-8 showed an 8-fold reduction in resistance to ceftazidime relative to FOX-3. In a kinetic analysis, FOX-8 displayed a 33-fold reduction in kcat/Km for ceftazidime compared to FOX-3. In the FOX family of β-lactamases, the Phe313 residue located in the R2 loop affects ceftazidime hydrolysis and alters the phenotype of E. coli strains carrying this variant.
Product Description
Ceftazidime for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ceftazidime pentahydrate, United States Pharmacopeia (USP) Reference Standard
Ceftazidime, European Pharmacopoeia (EP) Reference Standard