Merck
  • Synthesis of 5-(2-,3- and 4-methoxyphenyl)-4H-1,2,4-triazole-3-thiol derivatives exhibiting anti-inflammatory activity.
Synthesis of 5-(2-,3- and 4-methoxyphenyl)-4H-1,2,4-triazole-3-thiol derivatives exhibiting anti-inflammatory activity.
Farmaco (Societa chimica italiana : 1989) (2004-04-15)
L Labanauskas, E Udrenaite, P Gaidelis, A Brukstus
ABSTRACT
New S-alkylated 5-(2-,3- and 4-methoxyphenyl)-4H-1,2,4-triazole-3-thiols (5a-c, 6a-c) and 5-(2-,3- and 4-methoxyphenyl)-4-phenyl-4H-1,2,4-triazole-3-thiols (7a-c, 8a-c, 9a-c) were synthesized by the alkylation of 3-(2-,3- and 4-methoxyphenyl)-4,5-dihydro-1H-1,2,4-triazole-5-thiones (3a-c) or 3-(2-,3- and 4-methoxyphenyl)-4-phenyl-4,5-dihydro-1H-1,2,4-triazole-5-thiones (4a-c) with 1-iodobutane or 1-(1,3-benzodioxol-5-yl)-2-bromo-1-ethanone, 2-bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)-1-ethanone and 2-bromo-1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)-1-ethanone. Compounds 3a-c and 4a-c were synthesized by the acylation of thiosemicarbazide or 4-phenyl-3-thiosemicarbazide with 2-, 3- and 4-methoxybenzoyl chlorides and further cyclization of the obtained acylderivatives 1a-c and 2a-c. The synthesized compounds 4a-c, 5a, 6a-c, 7a-c, 8a-c, 9b,c exhibit anti-inflammatory activity.
Sigma-Aldrich
1-Iodobutane, 99%, contains copper as stabilizer