Merck
  • The PDE5 inhibitor vardenafil does not affect auditory sensory gating in rats and humans.
The PDE5 inhibitor vardenafil does not affect auditory sensory gating in rats and humans.
Psychopharmacology (2012-08-03)
O A H Reneerkens, A Sambeth, M A Van Duinen, A Blokland, H W M Steinbusch, J Prickaerts
ABSTRACT
Sensory gating is an adaptive mechanism of the brain to prevent overstimulation. Patients suffering from clinical disorders such as Alzheimer's disease or schizophrenia exhibit a deficit in gating, which indicates not only an impairment in basic information processing that might contribute to the cognitive problems seen in these patients. Phosphodiesterase type 5 inhibitors (PDE5-Is) have been shown to improve cognition in rodents in various behavioural tasks and might consequently be an interesting target for cognition enhancement. However, the effects of PDE5-Is on sensory gating are not known yet. This work aims to study the effects of PDE5 inhibition on auditory sensory gating in rats and humans. In the rat study, vehicle or 0.3-3 mg/kg of the PDE5-I vardenafil was given orally 30 min before testing and electrode locations were the vertex, hippocampus and the striatum. The human subjects received placebo, 10-20 mg vardenafil 85 min before testing and sensory gating was measured at the cortex (Fz, Fcz and Cz) electrodes. Significant gating was only found for the N1 component in rats while all three peaks P1, N1 and P2 showed gating in humans, i.e. the response to the second sound click was decreased as compared with the first for these deflections. Administration of vardenafil did neither have an effect on sensory gating in rats nor in humans. These findings imply that positive effects of PDE5 inhibition on cognition are not mediated by more early phases of information processing.
Product Description
Supelco
Zopiclone-D4 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Vardenafil dihydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®