Merck
  • Rapid and robust confirmation and quantification of 11-nor-Δ9-tetrahydrocannabinol-9-carboxylic acid (THC-COOH) in urine by column switching LC-MS-MS analysis.
Rapid and robust confirmation and quantification of 11-nor-Δ9-tetrahydrocannabinol-9-carboxylic acid (THC-COOH) in urine by column switching LC-MS-MS analysis.
Journal of mass spectrometry : JMS (2012-01-28)
Gabriele Zanchetti, Ivan Floris, Alberto Piccinotti, Silvia Tameni, Aldo Polettini
ABSTRACT
A method for the rapid and robust confirmation of 11-nor-∆9-tetrahydrocannabinol-9-carboxylic acid (THCA) in urine involving basic hydrolysis with NaOH and direct injection of the hydrolysate in a column-switching LC-MS-MS system was developed and validated. THCA-d3 was used as internal standard. Detection was performed in negative-ion mode by monitoring the transitions from the [M-CO(2) ]- ion m/z 299.2→245.2 and and m/z 299.2→191.1 that were found to provide a better signal-to-noise ratio than the transition from the pseudomolecular ion at m/z 343. The high sensitivity of detection enabled the injection of a small volume (10 µl) of the NaOH hydrolysate which, together with the applied column switching system, proved to confer ruggedness to the method and to avoid the deterioration of the instrumental apparatus despite the large amount of inorganic ions in the hydrolysate. The LLOQ was established at 5 ng/ml, and the LLOD was calculated as 0.2 ng/ml (S/N =3). The method was submitted to thorough validation including evaluation of the calibration range (5-500 ng/ml), accuracy and precision, matrix effects, overall process efficiency, autosampler stability, carryover and cross-talk, and 10-times reduction of sample volume (0.1 ml). Proof of applicability was obtained by direct comparison with the reference GC-MS method in use in the lab (the R(2) between the two methods was 0.9951).
Product Description
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
SKU
Pack Size
Availability
Price
Quantity
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®