Merck
  • [Plasma concentrations of 5-fluorouracil and F-beta-alanine following oral administration of S-1, a dihydropyrimidine dehydrogenase inhibitory fluoropyrimidine, as compared with protracted venous infusion of 5-fluorouracil].
[Plasma concentrations of 5-fluorouracil and F-beta-alanine following oral administration of S-1, a dihydropyrimidine dehydrogenase inhibitory fluoropyrimidine, as compared with protracted venous infusion of 5-fluorouracil].
Gan to kagaku ryoho. Cancer & chemotherapy (2006-08-11)
Yasuhide Yamada
ABSTRACT
The pharmacokinetics and pharmacodynamics of oral S-1, a dihydropyrimidine dehydrogenase (DPD) inhibitory fluoropyrimidine, were compared with those of protracted venous infusion (PVI) of 5-fluorouracil (5-FU). In all, 10 patients with gastric cancer received PVI of 5-FU at a dose of 250 mg/m2/day for 5 days. After a washout period of 9 days, the 10 patients received two divided doses daily for 28 days. S-1 was administered orally at about 9 a.m. and 7 p.m. Plasma concentrations of 5-FU and F-beta-alanine (FBAL) were measured for pharmacokinetic analysis, and the plasma uracil concentration was monitored as a surrogate marker of DPD inhibition in the same 10 patients on days 1-5 of PVI of 5-FU and on days 1-5 of oral S-1. The area under the curve (AUC0-10h) of 5-FU on day 5 was 728 +/- 113 ng x hr/ml for PVI of 5-FU and 1,364 +/- 374 ng x hr/ml for S-1. The median 5-FU PVI: S-1 ratio of the AUC0-10h of 5-FU was 1.9. The AUC0-10h of FBAL on day 5 of PVI of 5-FU was 9,465 +/- 3,225 ng x hr/ml, AUC0-10h, as compared with 1,725 +/- 605 ng x hr/ml on day 5 of S-1 treatment. The AUC0-10h of uracil on day 5 was 252 +/- 60 ng x hr/ml with PVI of 5-FU and 12,582 +/- 3,060 ng x hr/ml with S-1. The AUC0-10h of FBAL was markedly lower and plasma uracil concentrations were significantly higher for S-1 than for PVI of 5-FU, clearly demonstrating the effect of DPD inhibition.
Sigma-Aldrich
3-Amino-2-fluoropropionic acid, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
100 μL
Please contact Customer Service for Availability
€491.00
To order products, please contact your local dealer.