Merck
  • Inhibition of acid-catalyzed depolymerization of cellulose with boric acid in non-aqueous acidic media.
Inhibition of acid-catalyzed depolymerization of cellulose with boric acid in non-aqueous acidic media.
Carbohydrate research (2007-11-30)
Haruo Kawamoto, Shinya Saito, Shiro Saka
ABSTRACT
Boric acid inhibited the acid-catalyzed depolymerization of cellulose in sulfolane, a non-aqueous medium, at high temperature. Formation of the dehydration products such as levoglucosenone, furfural, and 5-hydroxymethyl furfural were also effectively inhibited. Similar inhibition was observed for cellooligosaccharides and starch, although the glucosidic bonds in methyl glucopyranosides and methyl cellobioside were cleaved to form alpha-d-glucofuranose cyclic 1,2:3,5-bisborate.
Product Description
Supelco
Sulfolane, analytical standard
Sigma-Aldrich
Sulfolane, 99%