Merck
  • Collagenase increases shortening of human bronchial smooth muscle in vitro.
Collagenase increases shortening of human bronchial smooth muscle in vitro.
American journal of respiratory and critical care medicine (1995-11-01)
A M Bramley, C R Roberts, R R Schellenberg
ABSTRACT
The recent demonstration of increased shortening of asthmatic airway smooth muscle could result from increased contractility of the muscle itself or from a decreased load that must be overcome by the smooth muscle to shorten. To evaluate the role of smooth muscle-associated extracellular matrix in limiting smooth muscle responses, we investigated the effect of collagenase on the mechanical responses of human bronchial smooth muscle strips. Contractile responses of second- to fourth-generation bronchi were evoked by electrical field stimulation, and measurements of length and tension were made at preloads between 0 and 2.5 g. The passive tension, active isometric, and isotonic responses were obtained at each preload before and after 90 min of incubation with 20 U/ml collagenase. Shortening to 10(-4) M histamine was also measured. Collagenase treatment caused a significant decrease in passive tension, with the most pronounced change occurring below Lmax (optimal length for force generation). At optimal lengths for shortening, the degree of shortening, expressed as a percentage of starting length, increased significantly from 8.9 +/- 1.4% before to 13.8 +/- 2.9% after collagenase treatment (n = 7) (p < 0.02). Shortening to histamine also increased from 14.3 +/- 2.5% before to 23.5 +/- 5.3% after collagenase treatment (n = 7) (p < 0.02). These results suggest that degradation of the collagenous matrix surrounding muscle in the airway wall reduces the load on the muscle, allowing increased smooth muscle shortening.
Sigma-Aldrich
Histamine bisphosphate monohydrate
Sigma-Aldrich
Histamine bisphosphate monohydrate, ≥99.0% (NT)