- Hemoptysis and respiratory failure following sildenafil use for pulmonary hypertension.
Hemoptysis and respiratory failure following sildenafil use for pulmonary hypertension.
Tennessee medicine : journal of the Tennessee Medical Association (2013-04-03)
Souheil M Abdel Nour, Holly Abdel Nour, Jay B Mehta, Thomas M Roy, Ryland P Byrd
PMID23544289
ABSTRACT
Sildenafil, usually a well-tolerated drug traditionally used for erectile dysfunction (ED), was recently approved for pulmonary arterial hypertension. In the literature, there are few cases of hemoptysis following sildenafil use for ED; however, to our knowledge, we are reporting the first case of hemoptysis following sildenafil use for pulmonary hypertension. We are documenting a case of a 90-year-old male patient who was admitted to the intensive care unit with hemoptysis and respiratory failure two weeks after he was started on sildenafil.
Product Description
Supelco
Sildenafil solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®