Merck
  • (-) Epicatechin attenuates mitochondrial damage by enhancing mitochondrial multi-marker enzymes, adenosine triphosphate and lowering calcium in isoproterenol induced myocardial infarcted rats.
(-) Epicatechin attenuates mitochondrial damage by enhancing mitochondrial multi-marker enzymes, adenosine triphosphate and lowering calcium in isoproterenol induced myocardial infarcted rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-12-25)
P Stanely Mainzen Prince
ABSTRACT
Cardiac mitochondrial damage plays an important role in the pathology of myocardial infarction. The protective effects of (-) epicatechin on cardiac mitochondrial damage in isoproterenol induced myocardial infarction were evaluated in rats. Rats were pretreated with (-) epicatechin (20 mg/kg body weight) daily for a period of 21 days. After the pretreatment period, isoproterenol (100 mg/kg body weight) was injected subcutaneously into rats twice at an interval of 24 h to induce myocardial infarction. Isoproterenol induced myocardial infarcted rats showed a significant increase in the levels of cardiac diagnostic markers, heart mitochondrial lipid peroxidation, calcium, and a significant decrease in the activities/levels of heart mitochondrial glutathione peroxidase, glutathione reductase, reduced glutathione, isocitrate, succinate, malate, α-ketoglutarate and NADH-dehydrogenases, cytochrome-C-oxidase and adenosine triphosphate. (-) Epicatechin pretreatment showed significant protective effects on all the biochemical parameters evaluated. The in vitro study revealed the superoxide and hydroxyl radical scavenging activity of (-) epicatechin. The possible mechanisms for the beneficial effects of (-) epicatechin on cardiac mitochondria could be attributed to scavenging of free radicals, decreasing calcium, increasing multi-enzymes (antioxidant, tricarboxylic acid cycle and respiratory chain enzymes), reduced glutathione and adenosine triphosphate. Thus, (-) epicatechin attenuated mitochondrial damage in isoproterenol induced myocardial infarcted rats.
Sigma-Aldrich
DL-Malic acid, 99%
Sigma-Aldrich
D-(+)-Malic acid, unnatural form, ≥97.0% (T)
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Supelco
α-Ketoglutaric acid disodium salt hydrate, ≥95%
Supelco
D-Malic acid, analytical standard
Sigma-Aldrich
α-Ketoglutaric acid disodium salt dihydrate, ≥98.0% (dried material, NT)
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, BioUltra
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, ≥98% (titration)
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Sigma-Aldrich
L-(−)-Malic acid disodium salt, ≥95% (titration)
Sigma-Aldrich
α-Ketoglutaric acid potassium salt, ≥98% (enzymatic)
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)