Merck
  • Synthesis and biological evaluation of 1,4-naphthoquinones and quinoline-5,8-diones as antimalarial and schistosomicidal agents.
Synthesis and biological evaluation of 1,4-naphthoquinones and quinoline-5,8-diones as antimalarial and schistosomicidal agents.
Organic & biomolecular chemistry (2012-07-11)
Don Antoine Lanfranchi, Elena Cesar-Rodo, Benoît Bertrand, Hsin-Hung Huang, Latasha Day, Laure Johann, Mourad Elhabiri, Katja Becker, David L Williams, Elisabeth Davioud-Charvet
ABSTRACT
Improving the solubility of polysubstituted 1,4-naphthoquinone derivatives was achieved by introducing nitrogen in two different positions of the naphthoquinone core, at C-5 and at C-8 of menadione through a two-step, straightforward synthesis based on the regioselective hetero-Diels-Alder reaction. The antimalarial and the antischistosomal activities of these polysubstituted aza-1,4-naphthoquinone derivatives were evaluated and led to the selection of distinct compounds for antimalarial versus antischistosomal action. The Ag(II)-assisted oxidative radical decarboxylation of the phenyl acetic acids using AgNO(3) and ammonium peroxodisulfate was modified to generate the 3-picolinyl-menadione with improved pharmacokinetic parameters, high antimalarial effects and capacity to inhibit the formation of β-hematin.
Sigma-Aldrich
1,4-Naphthoquinone, 97%
Sigma-Aldrich
1,4-Naphthoquinone, purum, ≥96.5% (HPLC)