Merck
  • Effect of L-deprenyl treatment on electrical activity, Na+, K+ ATPase, and protein kinase C activities in hippocampal subfields (CA1 and CA3) of aged rat brain.
Effect of L-deprenyl treatment on electrical activity, Na+, K+ ATPase, and protein kinase C activities in hippocampal subfields (CA1 and CA3) of aged rat brain.
Indian journal of experimental biology (2012-06-08)
Rameshwar Singh, Monika Mishra, Sangeeta Singh, Deepak Sharma
ABSTRACT
L-deprenyl is considered to protect against age-related cognitive deficits by improving long-term learning/memory in the aged brain. The CA1 and CA3 hippocampal areas are the sites at which initial learning and memory processes occur. Chronic deprenyl treatment significantly augmented the basal electrical firing rate (multiple-unit action potentials), and Na+, K(+)-ATPase and protein kinase C activities of both CA1 and CA3 indicating that the drug increased the excitability of CA1 and CA3. The increase, however, was much greater in CA1 than in CA3 suggesting that deprenyl can improve longer-term learning in aged animals by its excitability-enhancing action in CA1. The drug also countered the ageing-related loss of hippocampal protein kinase C activity.
Sigma-Aldrich
R-(−)-Deprenyl hydrochloride, powder, ≥98% (HPLC)
Selegiline hydrochloride, European Pharmacopoeia (EP) Reference Standard