Merck
  • C16 ceramide is crucial for triacylglycerol-induced apoptosis in macrophages.
C16 ceramide is crucial for triacylglycerol-induced apoptosis in macrophages.
Cell death & disease (2012-03-16)
E Aflaki, P Doddapattar, B Radović, S Povoden, D Kolb, N Vujić, M Wegscheider, H Koefeler, T Hornemann, W F Graier, R Malli, F Madeo, D Kratky
ABSTRACT
Triacylglycerol (TG) accumulation caused by adipose triglyceride lipase (ATGL) deficiency or very low-density lipoprotein (VLDL) loading of wild-type (Wt) macrophages results in mitochondrial-mediated apoptosis. This phenotype is correlated to depletion of Ca(2+) from the endoplasmic reticulum (ER), an event known to induce the unfolded protein response (UPR). Here, we show that ER stress in TG-rich macrophages activates the UPR, resulting in increased abundance of the chaperone GRP78/BiP, the induction of pancreatic ER kinase-like ER kinase, phosphorylation and activation of eukaryotic translation initiation factor 2A, the translocation of activating transcription factor (ATF)4 and ATF6 to the nucleus and the induction of the cell death executor CCAAT/enhancer-binding protein homologous protein. C16:0 ceramide concentrations were increased in Atgl-/- and VLDL-loaded Wt macrophages. Overexpression of ceramide synthases was sufficient to induce mitochondrial apoptosis in Wt macrophages. In accordance, inhibition of ceramide synthases in Atgl-/- macrophages by fumonisin B1 (FB1) resulted in specific inhibition of C16:0 ceramide, whereas intracellular TG concentrations remained high. Although the UPR was still activated in Atgl-/- macrophages, FB1 treatment rescued Atgl-/- macrophages from mitochondrial dysfunction and programmed cell death. We conclude that C16:0 ceramide elicits apoptosis in Atgl-/- macrophages by activation of the mitochondrial apoptosis pathway.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ddit3
Sigma-Aldrich
N-Palmitoyl-D-sphingosine, ≥98.0% (TLC)