Merck
  • High performance of N-alkoxycarbonyl-imines in triethylborane-mediated tin-free radical addition.
High performance of N-alkoxycarbonyl-imines in triethylborane-mediated tin-free radical addition.
The Journal of organic chemistry (2012-01-03)
Ken-ichi Yamada, Takehito Konishi, Mayu Nakano, Shintaro Fujii, Romain Cadou, Yasutomo Yamamoto, Kiyoshi Tomioka
ABSTRACT
Triethylborane-mediated tin-free radical alkylation of N-alkoxycarbonyl-imines, such as N-Boc-, N-Cbz-, and N-Teoc-imines, proceeded smoothly at a low temperature (-78 to -20 °C) to give the corresponding adducts in high yield. Although the formation of isocyanate was the major unfavorable reaction at room temperature, a one-pot conversion of N-Boc-imine to N-ethoxycarbonyl-adduct was possible through the corresponding isocyanate generated in situ. The higher performance of N-alkoxycarbonyl-imine than those of N-Ts- and N-PMP-imines is rationalized by a moderate electron-withdrawing character of an alkoxycarbonyl group that makes both addition of alkyl radical and trapping of the resulting aminyl radical by triethylborane efficiently fast.
Sigma-Aldrich
Triethylborane solution, 1.0 M in THF
Sigma-Aldrich
Triethylborane solution, 1.0 M in hexanes