Merck
  • Structural investigation of rimantadine inhibition of the AM2-BM2 chimera channel of influenza viruses.
Structural investigation of rimantadine inhibition of the AM2-BM2 chimera channel of influenza viruses.
Structure (London, England : 1993) (2011-11-15)
Rafal M Pielak, Kirill Oxenoid, James J Chou
ABSTRACT
The M2 channel of influenza A is a target of the adamantane family antiviral drugs. Two different drug-binding sites have been reported: one inside the pore, and the other is a lipid-facing pocket. A previous study showed that a chimera of M2 variants from influenza A and B that contains only the pore-binding site is sensitive to amantadine inhibition, suggesting that the primary site of inhibition is inside the pore. To obtain atomic details of channel-drug interaction, we determined the structures of the chimeric channel with and without rimantadine. Inside the channel and near the N-terminal end, methyl groups of Val27 and Ala30 from four subunits form a hydrophobic pocket around the adamantane, and the drug amino group appears to be in polar contact with the backbone oxygen of Ala30. The structures also reveal differences between the drug-bound and -unbound states of the channel that can explain drug resistance.
Sigma-Aldrich
1-(1-Adamantyl)ethylamine hydrochloride, 99%