Merck
  • A cationic water-soluble pillar[5]arene: synthesis and host-guest complexation with sodium 1-octanesulfonate.
A cationic water-soluble pillar[5]arene: synthesis and host-guest complexation with sodium 1-octanesulfonate.
Chemical communications (Cambridge, England) (2011-10-21)
Yingjie Ma, Xiaofan Ji, Fei Xiang, Xiaodong Chi, Chengyou Han, Jiuming He, Zeper Abliz, Weixiang Chen, Feihe Huang
ABSTRACT
By the introduction of trimethylammonium groups at both upper and lower rims, a cationic water-soluble pillar[5]arene was prepared, which forms a stable 1 : 1 host-guest complex with sodium 1-octanesulfonate in water.
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, free-flowing, Redi-Dri, ≥98%